Mouse alpha-1-antitrypsin ELISA Kit
Catalog # ABCAAB267809-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse alpha-1-antitrypsin ELISA kit
Specifications
About this item
Mouse alpha-1-antitrypsin ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse alpha-1-antitrypsin with a sensitivity of 65 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.