Human CSF-1-R ELISA Kit
ABCAAB230940-96
: Abcam
:
- Description:Human CSF-1-R ELISA kit
- Tests per kit:96 tests
Human CSF-1-R ELISA kit is a single-wash 90-min Simplestep used to quantify Human CSF-1-R with a sensitivity of 20,1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.