Rat GM-CSF ELISA Kit
Catalog # ABCAAB236709-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:GM-CSF
- Description:Rat GM-CSF ELISA kit
- Sample type:Serum, tissue culture media
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Detection range:3,1 - 250 pg/ml
- Storage temperature:4 °C
- Sensitivity:1,1 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Rat GM-CSF ELISA kit is a single-wash 90-min Simplestep used to quantify Rat GM-CSF with a sensitivity of 1.1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.