Rat Prolactin Receptor / PRL-R ELISA Kit
Catalog # ABCAAB238259-96
Supplier: Abcam
CAS Number:
Specifications
- Leírás:Rat Prolactin Receptor / PRL-R ELISA Kit
- Teszt/készlet:96 tests
Specifications
About this item
Rat Prolactin Receptor / PRL-R ELISA Kit is a single-wash 90-min Simplestep used to quantify Rat Prolactin Receptor / PRL-R with a sensitivity of 3,3 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.