Monkey GM-CSF ELISA Kit
ABCAAB252361-1X96
: Abcam
:
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:CD116
- Description:Monkey GM-CSF ELISA kit
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Detection range:15,63 - 1000 pg/ml
- Storage temperature:4 °C
- Sensitivity:8,45 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Monkey GM-CSF ELISA kit is a single-wash 90-min Simplestep used to quantify Monkey GM-CSF with a sensitivity of 8.45 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.