Rat PAI1 ELISA Kit (SERPINE1)
Supplier: Abcam
Certificates
About this item
Rat PAI1 ELISA kit (SERPINE1) is a single-wash 90-min Simplestep used to quantify Rat PAI1 (SERPINE1) with a sensitivity of 159 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders.
Frequently Bought Together