Mouse IP-10 ELISA Kit, Fluorescent
Catalog # ABCAAB272186-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse IP-10 ELISA kit, Fluorescent
Specifications
About this item
Mouse IP-10 ELISA kit, Fluorescent is a single-wash 90-min Simplestep used to quantify Mouse IP-10 with a sensitivity of 50,32 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Fluorescent Sandwich ELISA - 530/570/590 nm readout: works on any standard plate reader.