Mouse VCAM1 ELISA Kit (CD106)
Catalog # ABCAAB201278-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse VCAM1 ELISA Kit (CD106)
- Tests per kit:96 tests
Specifications
About this item
Mouse VCAM1 ELISA Kit (CD106) is a single-wash 90-min Simplestep used to quantify Mouse VCAM1 (CD106) with a sensitivity of 5,59 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.