Mouse NT-ProANP ELISA Kits
: Abcam
Mouse NT-ProANP ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse NT-ProANP with a sensitivity of 43,4 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.
Frequently Bought Together