Human Complement factor H ELISA Kits
Supplier: Abcam
Certificates
About this item
Human Complement factor H ELISA kit is a single-wash 90-min Simplestep used to quantify Human Complement factor H with a sensitivity of 74 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.
Specifications
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:Factor H
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Detection range:156,25 - 10000 pg/ml
- Storage temperature:4 °C
- Sensitivity:74 pg/ml
- Regulatory status:RUO
Specifications
Frequently Bought Together