Protein concentrator, Tube-O-CONCENTRATOR™
Supplier: G-Biosciences
Certificates
About this item
Tube-O-CONCENTRATOR™ Mini for 20 to 250 μl volumes or Medi for 0,2 to 2,5 ml volumes.
- Rapid concentration of protein solutions without protein precipitation
- Combination of a unique tube format dialysis device and a water absorbing liquid polymer
- Suitable for up to 250 µl or 2,5 ml protein solutions
- Zero protein loss, even if protein precipitation occurs
Tube-O-CONCENTRATOR™ is a versatile concentration device that utilizes a novel, water absorbing, liquid polymer and our patented Tube-O-DIALYZER™ for the rapid concentration of dilute protein solutions with zero protein loss. The unique tube design of Tube-O-DIALYZER™ ensures that 100% sample is recovered; simple place the entire device in a bench top centrifuge and spin for a few seconds.
Frequently Bought Together