Tubing clamps, Quickfit®
Supplier: SciLabware
Certificates
About this item
Tubing clamps made of thermoplastic polyester, for use with flexible rubber and plastic tubing.
- Provides fine flow control of liquids and gases
- Easy one-handed adjustment: just roll serrated wheel along sloped floor
Frequently Bought Together