(1S,2S)-(+)-Pseudoephedrine hydrochloride-[N-methyl-D3] 1000 µg/ml in methanol, Cerilliant
Catalog # PROOCERP-056
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:284665-25-4
- Description:(1S,2S)-(+)-Pseudoephedrine hydrochloride-[N-methyl-D3] 1,000 µg/ml in methanol
- Formula:C₁₀H₁₆ClNO
- Description:(1S,2S)-(+)-Pseudoephedrine hydrochloride-[N-methyl-D3]