Ground joint adaptors, BOLA
Supplier: Bohlender
Certificates
About this item
PTFE, white. For connecting different ground joint sizes.
- Temperature resistance: –200 to +250 °C
- Universal chemical resistance
- Autoclavable
Socket in cone, with sealing rings on the outside and knurled grip.
Frequently Bought Together