Bottle top burettes
About this item
The continuous bottle-top burette enables continuous titration, which leads to rapid, convenient, and accurate results.
- Long operating life from two easily-replaced micro-batteries 1,5 V
- Large, easy-to-read digits
- Simple, media-specific readjustment via keypad
- Continuous and precise regulation of titration speed with a large practical thumbwheel
- Reduced titrant waste with innovative recirculation valve
- Adaptable to bottles of different sizes
- Telescopic discharge tube which can be adjusted in both height and length
- Telescopic, extractable intake tube adapts burette to bottles of varying heights
- Continual supply of titration medium with specially developed double-piston pump
The angled display shows 4-position titration volume in large, easily read numbers, which simplifies operation. Turning the
two hand wheels supplies the titration medium in a continuous and pulse-free manner via the specially developed double-piston pump. Filling procedures are not necessary. This innovative technology increases safety; its compact design and low centre of gravity reduce risk of overturning, especially with smaller bottles. The height and length of the discharge tube can be adjusted, making it possible to work safely with both short and tall bottles. The innovative recirculation system prevents the loss of valuable reagent and reduces the risk of splashes. With its simple-to-use calibration function, continuous fulfils the corresponding requirements for test equipment monitoring without instrument downtime. Margins of error are under those specified in the DIN EN ISO 8655-3 standard, even for partial volumes. VITLAB® continuous is DE-M marked. Also available with DAkkS calibration certificate.
Certifications: DIN 12600, DIN EN ISO 8655-3
Delivery information: Supplied with thread adapters PP GL 32, GL 38 and S*40, a telescopic intake tube (200 to 350 mm), a telescopic discharge tube (140 to 220 mm), two micro-batteries of 1,5 volts (LR 03/AAA), and a quality certificate.
2 Options Available Below
Product Details & Documents
The continuous bottle-top burette enables continuous titration, which leads to rapid, convenient, and accurate results.
- Long operating life from two easily-replaced micro-batteries 1,5 V
- Large, easy-to-read digits
- Simple, media-specific readjustment via keypad
- Continuous and precise regulation of titration speed with a large practical thumbwheel
- Reduced titrant waste with innovative recirculation valve
- Adaptable to bottles of different sizes
- Telescopic discharge tube which can be adjusted in both height and length
- Telescopic, extractable intake tube adapts burette to bottles of varying heights
- Continual supply of titration medium with specially developed double-piston pump
The angled display shows 4-position titration volume in large, easily read numbers, which simplifies operation. Turning the
two hand wheels supplies the titration medium in a continuous and pulse-free manner via the specially developed double-piston pump. Filling procedures are not necessary. This innovative technology increases safety; its compact design and low centre of gravity reduce risk of overturning, especially with smaller bottles. The height and length of the discharge tube can be adjusted, making it possible to work safely with both short and tall bottles. The innovative recirculation system prevents the loss of valuable reagent and reduces the risk of splashes. With its simple-to-use calibration function, continuous fulfils the corresponding requirements for test equipment monitoring without instrument downtime. Margins of error are under those specified in the DIN EN ISO 8655-3 standard, even for partial volumes. VITLAB® continuous is DE-M marked. Also available with DAkkS calibration certificate.
Certifications: DIN 12600, DIN EN ISO 8655-3
Delivery information: Supplied with thread adapters PP GL 32, GL 38 and S*40, a telescopic intake tube (200 to 350 mm), a telescopic discharge tube (140 to 220 mm), two micro-batteries of 1,5 volts (LR 03/AAA), and a quality certificate.
Product Family Options
Product Information
Accessories
Product Information
- Catalog #LengthAvailabilityPrice
- Telescopic intake tube, FEP, ETFE, PTFETelescopic intake tube, FEP, ETFE, PTFEVITL1671085200 - 350 mmLog in for availability1 * 1 DB - 16 700 Ft