Wash bottle, LaboPlast®
Supplier: BURKLE
Certificates
About this item
LDPE, translucent.
- Immersion tube can be pulled up
- Good chemical resistance
- Good temperature resistance
The swan-neck jet produces a fine, straight spray with no lateral spatter. Dangerous lateral spatter does not occur.
Frequently Bought Together