HEPPSO (β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid)
Supplier: Molekula
Certificates
About this item
CAS Number:68399-78-0
:
Formula:C₉H₂₀N₂O₅S
MDL Number:MFCD00038351
:268.3305 g/mol
EINECS:269-990-1
:GHS08
Storage Temperature:Ambient
Frequently Bought Together