(1S)-(-)-Camphanic chloride
Catalog # MOLEM42033749
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:216.67 g/mol
EINECS:254-552-4
UN:3261
ADR:8,II
CAS Number:39637-74-6
Formula:C₁₀H₁₃ClO₃
MDL Number:MFCD00135626