Sterilin® Dippas® Milk Dippers
Supplier: STERILIN
Certificates
About this item
These sample dippers facilitate the sample collection and subsequent transportation to the laboratory in a single container with no risk of cross contamination.
- Individually wrapped and gamma irradiated to ensure sterility
- Available in either clear or blue PS for the food industry
- Handle neatly snaps off after sample collection for ease of transportation
Certifications: EN 14245 and BS5213.
Delivery: Supplied with a 'flow-seal' screw cap for a leak tight seal.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Frequently Bought Together