Order Entry
Hungary
ContactUsLinkComponent
2'-Deoxyuridine
2'-Deoxyuridine
Catalog # MOLEM85585786
Supplier:  Molekula
CAS Number:  
2'-Deoxyuridine
Catalog # MOLEM85585786
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :228.20 g/mol
Storage Temperature:Ambient
EINECS:213-455-7
CAS Number:951-78-0
Formula:C₉H₁₂N₂O₅
MDL Number:MFCD00006527