Order Entry
Hungary
ContactUsLinkComponent
Magnetic stirrers, battery powered, SP Bel-Art
Magnetic stirrers, battery powered, SP Bel-Art
Catalog # 442-9460
Magnetic stirrers, battery powered, SP Bel-Art
Catalog # 442-9460

Specifications

  • Description:
    Magnetic stirrer, battery powered
  • Model:
    Battery powered
  • Speed range:
    900 min⁻¹
  • W×D×H:
    235×132×89 mm
  • Max. stirring capacity H₂O:
    1000 ml
  • Plate material:
    PP
  • Pk:
    1

Specifications

About this item

This portable magnetic stirrer is made of PP.

  • Durable and chemical resistant
  • Two D batteries provide 16 days of continuous operation

Ideal for use inside incubators and when electricity is not available.

Delivery: Supplied with Spinbar® Octagon Stirbar (BELAF371100138).