Drying tubes with tube fittings, SP Bel-Art
Gyártó: Bel-Art Products, a Part of SP
Tanúsítványok
Termék információk
Unbreakable polyethylene straight tubes.
- Good chemical resistance
- Godd temperature resistance
- Drying agent no included
Can be filled with desiccant of your choice. Each tube is supplied with two polypropylene fittings which accept 6,4 to 9,5 mm int.Ø tubing.
Frequently Bought Together