Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Regisztráció
Products
Services
Digital Solutions
Applications
Support
About
Rendelés felvitel
Megrendelések állapota
Saját fiók
Kosár
Hungary
Rendelés felvitel
Megrendelések állapota
Saját fiók
Kosár
ContactUsLinkComponent
Home
Clamps
General Purpose Clamps
Clips for safe bags, Clavies®, SP Bel-Art
Clips for safe bags, Clavies®, SP Bel-Art
Clips for safe bags, Clavies®, SP Bel-Art
Catalog # BELAF131900100
Supplier:
Bel-Art Products, a Part of SP
Expand image
Clips for safe bags, Clavies®, SP Bel-Art
Catalog # BELAF131900100
Supplier:
Bel-Art Products, a Part of SP
Specifications
For:
autoclavable bags from 304×609 to 482×584 mm
Colour:
Orange
Description:
Safe bag clips
Pk:
100
Specifications
About this item
Product Details
Product Details