Order Entry
Hungary
ContactUsLinkComponent
Tin standard solution, 1000 mg/L Sn in 1 M hydrochloric acid standard for AAS
Tin standard solution, 1000 mg/L Sn in 1 M hydrochloric acid standard for AAS
Catalog # REGEAASNH
Supplier:  REAGECON
CAS Number:  
Tin standard solution, 1000 mg/L Sn in 1 M hydrochloric acid standard for AAS
Catalog # REGEAASNH
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    1 M HCl
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Tin
  • Symbol:
    Sn

Specifications

About this item