Potassium standard solution, 1,000 mg/L K in 0.5 M nitric acid (from KNO₃) standard for AAS
Catalog # SCARPO01050500
Supplier: Scharlau
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Glass bottle
- Pk:500 mL
- Element:Potassium
- Symbol:K