Order Entry
Hungary
ContactUsLinkComponent
Standard solution, phosphate, 500 mg/L as PO4 (NIST), pk/16 - 10-mL voluette® ampules
Standard solution, phosphate, 500 mg/L as PO4 (NIST), pk/16 - 10-mL voluette® ampules
Catalog # 705-0058
Supplier:  Hach
CAS Number:  
Standard solution, phosphate, 500 mg/L as PO4 (NIST), pk/16 - 10-mL voluette® ampules
Catalog # 705-0058
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    16
  • Description:
    Phosphate standard, 500 mg/l in 10 ml ampoules
  • Pk Info:
    16 ampoules
Specifications
About this item
For accuracy checks in phosphate (phosphorus) determinations
  • Simple Accuracy Checks
  • Minimize waste
  • Reliable results