Order Entry
Hungary
ContactUsLinkComponent
 
Corning® mouse laminin (from Engelbreth-Holm-Swarm mouse tumour)
  734-1098
 :  Corning
   Low Manufacturing Impact
 :  
undefined
Corning® mouse laminin (from Engelbreth-Holm-Swarm mouse tumour)
  734-1098
 :  Corning
   Low Manufacturing Impact
 :  

 

  • Pk:
    1 mg
  • Source:
    Engelbreth-Holm-Swarm mouse tumour
  • Species:
    Mouse
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Storage conditions:
    Stable for at least three months at −70 °C. Do not store in frost-free freezer. Avoid multiple freeze-thaws.
  • Environmental certification:
    ISO 14001 - Bedford
  • Protein/peptide name:
    Laminin
  • Formulation:
    1 mg/ml frozen in 0,05 M Tris-HCl

 

 

Laminin, a major component of basement membranes, has numerous biological activities including promotion of cell adhesion, migration, growth, and differentiation, including neurite outgrowth. It can be used as a thin coating on tissue culture surfaces or as a soluble additive to culture medium. The Laminin/Entactin Complex, high concentration (HC) is a special formulation that has been developed for three-dimensional (3D) culture.

Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma.