Blood gas capillary tubes, VITREX®
Supplier: Vitrex Medical A/S
Certificates
About this item
These disposable capillary tubes for blood gas analyses are made from heavy walled glass.
- Available non-heparinised, with sodium or lithium heparinisation
- Careful fire polishing, heavy walled tubes and robust end-caps secure proper function and reduce operating risk
- Both ends melted
Shelf life: 4 years.
Frequently Bought Together