Order Entry
Hungary
ContactUsLinkComponent
Corning® Mouse collagen IV (from Engelbreth-Holm-Swarm lathrytic mouse tumour)
Corning® Mouse collagen IV (from Engelbreth-Holm-Swarm lathrytic mouse tumour)
Catalog # 734-0099
Supplier:  Corning
CAS Number:  
undefined
Corning® Mouse collagen IV (from Engelbreth-Holm-Swarm lathrytic mouse tumour)
Catalog # 734-0099
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
    Engelbreth-Holm-Swarm lathrytic mouse tumour
  • Species:
    Mouse
  • Storage conditions:
    Stable for at least three months at −70 °C
  • Protein/peptide name:
    Collagen IV
  • Formulation:
    Frozen in 0,05 M HCl

Specifications

About this item

Collagen IV is a ubiquitous component of basement membranes, the sheet-like matrix that underlies epithelial and endothelial cells and surrounds muscle fat and nerve cells. It can be used as a thin coating on tissue culture surfaces to promote cell attachment and proliferation and to study its effects on cell behaviour.

Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma. Source material tested for hepatitis B antigen and HIV-1 antibody