Order Entry
Hungary
ContactUsLinkComponent
 
2-Nitrothiophene
Catalog # MOLEM50354243
Supplier:  Molekula
CAS Number:  
2-Nitrothiophene
Catalog # MOLEM50354243
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :129.14 g/mol
Boiling Pt:224 °C (1013 hPa)
Boiling Pt:224 °C (1013 hPa)
Melting Pt:46.5 °C
Melting Pt:46,5 °C
Density:1,3644 g/cm³ (42.6 °C)
Storage Temperature:Ambient
EINECS:210-190-9
CAS Number:609-40-5
Formula:C₄H₃NO₂S
MDL Number:MFCD00005425