Order Entry
Hungary
ContactUsLinkComponent
 
 
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
 
undefined
 
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
 

 

  • Assay:
    98.5 - 100.5 %
  • Appearance:
    White pastilles
  • Appearance of solution:
    Passes test Ph.Eur.
  • Identification B:
    Passes test Ph.Eur.
  • Solution in water:
    Passes test
  • pH (20°C; 0.01 %):
    Min. 11.0
  • Heavy metals (as Ag):
    Max. 20 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Total N (Nitrogen):
    Max. 3 ppm
  • CO3 (as Na2CO3):
    Max. 1.0 %
  • Cl (Chloride):
    Max. 120 ppm
  • PO4 (Phosphate):
    Max. 5 ppm
  • SiO4 (as SiO2):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 5 ppm
  • As (Arsenic):
    Max. 1 ppm
  • Al (Aluminium):
    Max. 5 ppm
  • Ca (Calcium):
    Max. 5 ppm
  • Cu (Copper):
    Max. 2 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Hg (Mercury):
    Max. 0.1 ppm
  • K (Potassium):
    Max. 0.1 %
  • Mg (Magnesium):
    Max. 5 ppm
  • Ni (Nickel):
    Max. 5 ppm
  • Pb (Lead):
    Max. 5 ppm
  • Zn (Zinc):
    Max. 10 ppm
  • Conforms to Reag. Ph.Eur.:
    Passes test
  • -:
    -

 

Frequently Bought Together