Magnetic Stirring Bars, glass, cylindrical, BOLA
Supplier: Bohlender
Certificates
About this item
Cylindrical magnetic stirring bars with Alnico V magnetic core encapsulated in borosilicate glass.
- Non porous, non contaminating, universal chemical resistance
- For high-purity work or trace analysis
Cylindrical shape with extremely smooth surface prevents penetration of substances.
Frequently Bought Together