
4-Quinolinol
Catalog # MOLE76234561-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:145.16 g/mol
Melting Pt:202…206 °C
EINECS:210-268-2
:GHS07
Storage Temperature:Ambient
CAS Number:611-36-9
Formula:C₉H₇NO
MDL Number:MFCD00006777