Order Entry
Hungary
Orders LinkContactUsLinkComponent
 

4,7-Dichloroquinoline

100 mg
Catalog # PROOMM0617.03
Irritant
4,7-Dichloroquinoline
Catalog # PROOMM0617.03
CAS Number:  86-98-6
4,7-Dichloroquinoline

100 mg

CAS Number:  86-98-6
Price
481 000 Ft
Log in for availability
1 * 100 mg
Product Details & Documents
  • CAS Number:86-98-6
  • Formula:C₉H₅Cl₂N
  • MDL Number:MFCD00006774
  • Molecular Weight:198.05 g/mol
  • Boiling Pt:293 °C (1013 hPa)
  • Melting Pt:81…83 °C
  • Storage Temperature:Ambient
  • EINECS:201-714-7
Specifications

Specifications

  • Pk:
    100 mg

Specifications