Benzoin
Catalog # MOLEM66437043
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:212.25 g/mol
EINECS:204-331-3
CAS Number:119-53-9
Formula:C₁₄H₁₂O₂
MDL Number:MFCD00004496