Order Entry
Hungary
ContactUsLinkComponent
 
 
 
 
 
n-Hexane ≥97%, HiPerSolv CHROMANORM® for preparative HPLC
 
undefined
 
 
 
 
n-Hexane ≥97%, HiPerSolv CHROMANORM® for preparative HPLC
 

 

  • Assay (calculated on anhydrous):
    Min. 97.0 %
  • Acidity:
    Max. 0.0003 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 50 ppm

 

Frequently Bought Together