Order Entry
Hungary
ContactUsLinkComponent
 
 
1,2-Dibromo-1-phenylethane 97%
  ACRO299305000
 :  
undefined
 
1,2-Dibromo-1-phenylethane 97%
  ACRO299305000
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Appearance:
    White to cream to yellow Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    70 to 75 °C
  • GC:
    ≥96.0 %

 

 

 :
 :263.96 g/mol
 :139…141 °C (15 mmHg)
 :68…73 °C
 :1,74 g/cm³ (20 °C)
 :Ambient
 :202-253-4
 :3261
 :8,II
 :GHS05
 :93-52-7
 :C₆H₅CHBrCH₂Br
 :MFCD00000138