Order Entry
Hungary
ContactUsLinkComponent
Retort ring for crucibles
Catalog # BOCH5505
Supplier:  BOCHEM
undefined
Retort ring for crucibles
Catalog # BOCH5505
Supplier:  BOCHEM
Specifications
  • Ø int.:
    0 - 70 mm
  • Description:
    Crucible retort ring
  • Pk:
    1
Specifications
About this item
Crucible retort ring made of stainless steel. Ideal for clamping of crucibles for heating substances over a gas flame.
  • Oriental clamp for crucibles, which can be fixed easily by hand according to the used crucible size: 0 to 70 mm
  • 3-point-clamp: ring with 3 guide sleeves pointing to the center, the clamping rods inside the guide sleeves can be scrolled
  • Clamping rods: ceramic rods 'Degussit' (max. 2030 °C)
  • Can be combined with support stands or tripods; with the support stands the distance between the gas flame and crucible bottom can be adjusted
  • Also suitable for platinum crucibles