Order Entry
Hungary
ContactUsLinkComponent
Pipette filler
Pipette filler
Catalog # KART198
Supplier:  KARTELL
undefined
Pipette filler
Catalog # KART198
Supplier:  KARTELL
Specifications
  • Capacity:
    10 ml
  • Description:
    Pipette filler with plunger, ABS/HDPE, with graduations
  • Pk:
    1
Specifications
About this item
Pipette filler with plunger, will handle pipettes up to 10 ml.
  • Comfortable grip
  • Simply press plunger and release to fill pipette
  • Knurled cap adjusts the plunger stroke for repeating pre-set volume
  • Rubber chuck will hold pipette securely and safely
  • Graduations on the body
  • Moulded in yellow ABS