Order Entry
Hungary
Orders LinkContactUsLinkComponent
Pen needles, BD Micro-Fine™

Pen needles, BD Micro-Fine™

Thin wall, single use, 31, 8 mm, 0,25 mm, Pen needle, Blue
Catalog # BDAM320631
Pen needles, BD Micro-Fine™
Catalog # BDAM320631
Supplier:  
Pen needles, BD Micro-Fine™

Thin wall, single use, 31, 8 mm, 0,25 mm, Pen needle, Blue

Discontinued
Product Details & Documents
Specifications
  • Colour code:
    Blue
  • Ø:
    0,25 mm
  • Gauge:
    31
  • Type:
    Pen needle
  • Description:
    Thin wall, single use
  • Length:
    8 mm