Order Entry
Hungary
ContactUsLinkComponent
 
Irritant
5-Iodoanthranilic acid
Catalog # MOLE50555063-25G
Supplier:  Molekula
CAS Number:  
Irritant
5-Iodoanthranilic acid
Catalog # MOLE50555063-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :263.03 g/mol
Boiling Pt:360 °C (1013 hPa)
Boiling Pt:360 °C (1013 hPa)
Melting Pt:234 °C
EINECS:226-205-7
 :GHS07
CAS Number:5326-47-6
Formula:C₇H₆INO₂
MDL Number:MFCD00007849