

2,4-Difluorobenzaldehyde
Catalog # MOLEM43683780
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:142.1 g/mol
Boiling Pt:82 °C (42 mmHg)
Density:1.299 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-287-2
UN:1989
ADR:3,III
:GHS02
:GHS07
CAS Number:1550-35-2
Formula:C₇H₄F₂O
MDL Number:MFCD00010326