DURAN® Adapters, Receiver, Vacuum, Straight
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Straight, with two standard ground joints and screw-on plastic hose connection.
Frequently Bought Together