Stirrer bearing with screw thread DURAN® KPG®
Gyártó: DWK Life Sciences
Tanúsítványok
Termék információk
DURAN®, clear borosilicate glass 3.3.
- Interchangeable
- With standard ground joint and GL screw thread tube
- Ground and polished surface
Frequently Bought Together