0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Heaters
Gas Burners
Blowpipes for Usbeck burners
Blowpipes for Usbeck burners
Blowpipes for Usbeck burners
Catalog # 451-2911
Supplier:
USBECK Laborgeräte
Expand image
Blowpipes for Usbeck burners
Catalog # 451-2911
Supplier:
USBECK Laborgeräte
Specifications
Description:
Blowpipe, with wooden mouth piece, nickel plated brass, length: 250 mm
Pk:
1
Specifications
About this item
Product Details
Product Details