Distilling receivers, Bredt, bent
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With four flasks NS 14/23, with bent adapter (105°) vacuum connection: GL 18.
Frequently Bought Together