Burettes, pellet pattern, lateral stopcock, Schellbach stripe
Supplier: TECHNOSKLO
Certificates
About this item
Borosilicate glass 3.3, class AS, conformity certified.
- Calibrated to deliver ’Ex’, scale length 500 mm
- Batch identification is printed on the burette
- Automatic zeroing
Delivery: Delivered without 2 L bottle nor burette bulb
Frequently Bought Together