Order Entry
Hungary
ContactUsLinkComponent
Beakers, tall form
Beakers, tall form
Catalog # 213-0333
Supplier:  TECHNOSKLO
Supplier #: 632.427012150
undefined
Beakers, tall form
Catalog # 213-0333
Supplier:  TECHNOSKLO
Supplier #: 632.427012150
Log in for availability
Select an Option
54 mm
Select an Option
150 ml
Select an Option
95 mm
Select an Option
03
Select an Option
10
About this item
Borosilicate glass 3.3, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
Frequently Bought Together