Order Entry
Hungary
ContactUsLinkComponent
Trays, disinfection
  216-0930
undefined
Trays, disinfection
  216-0930
  • W×D×H:
    250×165×60 mm
  • Pk:
    1
Glass.
  • Smooth surfaces for easy cleaning
  • Sturdy
  • Good chemical resistance
Multipurpose trays for a wide range of uses.