Funnels with short stem
Supplier: TECHNOSKLO
Certificates
About this item
Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With smooth internal walls. Angle 60°.
Frequently Bought Together